Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029860.3 | NP_001025031.2 | 2 - 3 | 2305 | 151 | |
| NR_036491.1 | 2 - 3 | 2349 | 151 | |||
| GenBank mRNA | BC090892.1 | - | 2 - 3 | 2278 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08019137.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019137.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019135.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019135.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019135.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019135.hg.1 | TC08001475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176587.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800176590.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176590.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176590.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800176590.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800176590.hg.1 | TC0800011243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000428847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000517806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | ENST00000520987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NM_001029860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011243.hg.1 | NR_036491.1 | Refseq |

FBXO43
Genomic Map