Assay Details
| Gene Symbol: ZBTB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020924.3 | NP_065975.1 | 1 - 2 | 118 | 86 | |
| GenBank mRNA | BC018748.1 | - | 1 - 2 | 67 | 4553653 | 86 |
| HY066289.1 | - | 1 - 2 | 119 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018748.1, NM_020924.3 transcripts.
Set Membership: |
|

ZBTB26
Genomic Map