| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009556.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170465.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170465.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170465.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170465.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170465.hg.1 |
TC1400009556.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400170464.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170464.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170464.hg.1 |
ENST00000599174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400170464.hg.1 |
NM_001271507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400170464.hg.1 |
TC1400009556.hg.1 |
Clariom D TC |