Assay Details
| Gene Symbol: DBET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121644.1 | 1 | 2401 | 60 | ||
| GenBank mRNA | JQ639078.1 | - | 3 | 2397 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DBET
Genomic Map