Assay Details
| Gene Symbol: CUL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324375.1 | NP_001311304.1 | 1 - 2 | 207 | 104 | |
| NM_003591.3 | NP_003582.2 | 1 - 2 | 207 | 104 | ||
| GenBank mRNA | AK294080.1 | - | 1 - 2 | 155 | 104 | |
| BC009591.1 | - | 1 - 2 | 190 | 4104375 | 104 | |
| DC401936.1 | - | 1 - 2 | 207 | 104 | ||
| U83410.1 | - | 1 - 2 | 126 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK294080.1, BC009591.1, NM_001324375.1, NM_003591.3, U83410.1 transcripts.
Set Membership: |
|

CUL2
Genomic Map