Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077619.1 | NP_001071087.1 | 5 - 6 | 657 | 124 | |
| GenBank mRNA | AK126300.1 | - | 5 - 6 | 560 | 124 | |
| AL831990.2 | - | 5 - 6 | 531 | 124 | ||
| AL833075.2 | - | 5 - 6 | 531 | 124 | ||
| BC113645.1 | - | 5 - 6 | 657 | 8322697 | 124 | |
| BX647876.2 | - | 5 - 6 | 531 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000402.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08005632.hg.1 | ENST00000399598.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005632.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08005632.hg.1 | TC08000402.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800153230.hg.1 | TC0800007735.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000399598.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000520732.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000521796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000522978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | ENST00000523409.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800007735.hg.1 | NM_001077619.1 | Refseq |

UBXN2B
Genomic Map