Assay Details
| Gene Symbol: PSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031246.3 | NP_112536.2 | 4 - 5 | 1132 | 78 | |
| GenBank mRNA | M20882.1 | - | 4 - 5 | 1043 | 78 | |
| M37398.1 | - | 1 - 2 | 233 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001581.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | ENST00000329509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | ENST00000401942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | ENST00000406487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | ENST00000406917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021653.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | ENST00000329509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | ENST00000401942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | ENST00000406487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | ENST00000406917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021652.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021650.hg.1 | ENST00000329509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021650.hg.1 | ENST00000401942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021650.hg.1 | ENST00000406487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021650.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021650.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | ENST00000329509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | ENST00000401942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | ENST00000406487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | ENST00000406917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021651.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | ENST00000329509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | ENST00000401942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | ENST00000406487.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | ENST00000406917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021655.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021658.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021658.hg.1 | TC19001581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011965.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206491.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206492.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206496.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206496.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206496.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206496.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206496.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206497.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206497.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206497.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206497.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206497.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206499.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | ENST00000491995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206500.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | ENST00000593482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206493.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000501199.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | ENST00000597171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011962.hg.1 | NR_026824.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | ENST00000329509.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | ENST00000406487.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206495.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206501.hg.1 | NM_031246.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206501.hg.1 | TC1900011965.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206501.hg.1 | NM_031246.3 | Refseq |

PSG2
Genomic Map