Assay Details
| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012342.2 | NP_036474.1 | 1 - 2 | 480 | 52 | |
| GenBank mRNA | AK223065.1 | - | 1 - 2 | 452 | 52 | |
| BC019252.1 | - | 1 - 2 | 465 | 3503549 | 52 | |
| BT006820.1 | - | 1 - 2 | 77 | 52 | ||
| U23070.1 | - | 1 - 2 | 449 | 52 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000199.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transforming growth factor beta receptor signaling pathway
positive regulation of cell proliferation
regulation of cell shape
positive regulation of epithelial to mesenchymal transition
cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of protein binding
positive regulation of catenin import into nucleus
positive regulation of transcription, DNA-templated
positive regulation of canonical Wnt signaling pathway
frizzled binding
type II transforming growth factor beta receptor binding
transforming growth factor beta receptor signaling pathway
positive regulation of cell proliferation
regulation of cell shape
positive regulation of epithelial to mesenchymal transition
cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of protein binding
positive regulation of catenin import into nucleus
positive regulation of transcription, DNA-templated
positive regulation of canonical Wnt signaling pathway
frizzled binding
type II transforming growth factor beta receptor binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10002387.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002387.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002387.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002387.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002384.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002384.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002384.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002384.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002385.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002385.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002385.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002385.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002391.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002391.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002391.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002391.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002394.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002394.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002394.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002392.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002392.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002392.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002392.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002393.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002393.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002393.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002393.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002386.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002386.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002386.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002389.hg.1 | ENST00000375533.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002389.hg.1 | ENST00000542444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002389.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10002389.hg.1 | TC10000199.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148632.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007132.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148633.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148635.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148637.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148637.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148637.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148637.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148637.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148636.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | ENST00000497699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148631.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148638.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148638.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148638.hg.1 | ENST00000375533.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148638.hg.1 | NM_012342.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148638.hg.1 | TC1000007132.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 155488 | - | - |

BAMBI
Genomic Map