Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031721.3 | NP_001026891.2 | 5 - 6 | 696 | 122 | |
| NM_024840.3 | NP_079116.2 | 5 - 6 | 588 | 122 | ||
| XM_005259269.3 | XP_005259326.1 | 4 - 5 | 802 | 122 | ||
| XM_011527333.2 | XP_011525635.1 | 5 - 6 | 912 | 122 | ||
| XM_017027315.1 | XP_016882804.1 | 4 - 5 | 760 | 122 | ||
| GenBank mRNA | AK027565.1 | - | 5 - 6 | 588 | 122 | |
| BC057776.1 | - | 5 - 6 | 655 | 30338140 | 122 | |
More Information
Set Membership: |
|

ZNF613
Genomic Map