Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000531.5 | NP_000522.3 | 1 - 2 | 295 | 70 | |
| XM_017029556.1 | XP_016885045.1 | 1 - 2 | 250 | 70 | ||
| GenBank mRNA | AK292757.1 | - | 1 - 2 | 250 | 70 | |
| BC074745.2 | - | 1 - 2 | 117 | 30915205 | 70 | |
| BC107153.1 | - | 1 - 2 | 295 | 40032502 | 70 | |
| BC107154.1 | - | 1 - 2 | 295 | 40032511 | 70 | |
| BC114496.1 | - | 1 - 2 | 294 | 40083569 | 70 | |
| CB158478.1 | - | 1 - 2 | 201 | 70 | ||
| K02100.1 | - | 1 - 2 | 216 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000177.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
urea cycle
liver development
ornithine catabolic process
midgut development
response to zinc ion
citrulline biosynthetic process
response to insulin
arginine biosynthetic process via ornithine
response to drug
anion homeostasis
protein homotrimerization
response to biotin
ammonia homeostasis
ornithine carbamoyltransferase activity
phospholipid binding
amino acid binding
phosphate ion binding
urea cycle
liver development
ornithine catabolic process
midgut development
response to zinc ion
citrulline biosynthetic process
response to insulin
arginine biosynthetic process via ornithine
response to drug
anion homeostasis
protein homotrimerization
response to biotin
ammonia homeostasis
ornithine carbamoyltransferase activity
phospholipid binding
amino acid binding
phosphate ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002035.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002035.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002035.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002035.hg.1 | TC0X000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002032.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002032.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002032.hg.1 | TC0X000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002033.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002033.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002033.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002033.hg.1 | TC0X000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002034.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002034.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002034.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002034.hg.1 | TC0X000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002031.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002031.hg.1 | TC0X000177.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147478.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147478.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147478.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147478.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147478.hg.1 | TC0X00006970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147482.hg.1 | TC0X00006970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147477.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147477.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147477.hg.1 | TC0X00006970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147481.hg.1 | TC0X00006970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147480.hg.1 | TC0X00006970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006970.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | ENST00000039007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | ENST00000488812.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | NM_000531.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147479.hg.1 | TC0X00006970.hg.1 | Clariom D TC |

OTC
Genomic Map