| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X002036.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X002036.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X002036.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X002036.hg.1 |
TC0X000177.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006970.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
ENST00000039007.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
ENST00000488812.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
NM_000531.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147483.hg.1 |
TC0X00006970.hg.1 |
Clariom D TC |