Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000531.5 | NP_000522.3 | 4 - 5 | 599 | 116 | |
| XM_017029556.1 | XP_016885045.1 | 4 - 5 | 554 | 116 | ||
| GenBank mRNA | AK292757.1 | - | 4 - 5 | 554 | 116 | |
| BC074745.2 | - | 4 - 5 | 421 | 30915205 | 116 | |
| BC107153.1 | - | 4 - 5 | 599 | 40032502 | 116 | |
| BC107154.1 | - | 4 - 5 | 599 | 40032511 | 116 | |
| BC114496.1 | - | 4 - 5 | 598 | 40083569 | 116 | |
More Information
Set Membership: |
|

OTC
Genomic Map