| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04021352.hg.1 |
ENST00000296509.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04021352.hg.1 |
NM_002358.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04021352.hg.1 |
TC04001507.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000296509.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000333047.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000504707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000512484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
NM_002358.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000296509.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000333047.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000504707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
ENST00000512484.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
NM_002358.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400180570.hg.1 |
TC0400011721.hg.1 |
Clariom D TC |