Assay Details
| Gene Symbol: RMRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003051.3 | 1 | 147 | 102 | ||
| GenBank mRNA | AA593275.1 | - | 1 | 122 | 101 | |
| DW422801.1 | - | 1 | 163 | 102 | ||
| DW423815.1 | - | 1 | 163 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3401284 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4082 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4081 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56412 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57559 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3401284 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00318271 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3401284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16174554 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16174553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16174554 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16174553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16174554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16174547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699360 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699385 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699376 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06699377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16174566 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003051_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161025 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8161024 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014014.hg.1 | ENST00000363046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014014.hg.1 | NR_003051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014014.hg.1 | TC09001056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC09001056.hg.1 | NR_003051.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC09001056.hg.1 | ENST00000363046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | ENST00000363046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | ENST00000602361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | NR_003051.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | ENST00000363046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | ENST00000602361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | NR_003051.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168378.hg.1 | TC0900009947.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009947.hg.1 | ENST00000363046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009947.hg.1 | ENST00000602361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009947.hg.1 | NR_003051.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037309 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3242405 | NR_003051.3 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242405 | AA593275.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242405 | DW422801.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242405 | DW423815.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3242405 | NR_003051.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3242405 | AA593275.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3242405 | DW422801.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3242405 | DW423815.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242405 | NR_003051.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242405 | AA593275.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242405 | DW422801.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242405 | DW423815.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242405 | NR_003051.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242405 | AA593275.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242405 | DW422801.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242405 | DW423815.1 | human_transcriptome |

RMRP
Genomic Map