Assay Details
| Gene Symbol: SNORA24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002963.1 | 1 | 61 | 77 | ||
| GenBank mRNA | AJ609460.1 | - | 1 | 61 | 77 | |
| AJ609478.1 | - | 1 | 61 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000600.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3963 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3962 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15605698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15605698 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03382601 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P498767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03382598 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03382601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002963_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8097057 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8097056 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008712.hg.1 | ENST00000384096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008712.hg.1 | NR_002963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008712.hg.1 | TC04000600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012351.hg.1 | ENST00000384401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000384096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_002963.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009792.hg.1 | ENST00000384176.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | ENST00000384096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | NR_002963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | ENST00000384096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | NR_002963.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158523.hg.1 | TC0400008537.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000384096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | ENST00000602819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_002963.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_003584.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008537.hg.1 | NR_034011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012351.hg.1 | ENST00000384401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009792.hg.1 | ENST00000384176.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3248890 | NR_002963.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3248890 | AJ609460.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248890 | NR_002963.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248890 | AJ609460.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248890 | NR_002963.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248890 | AJ609460.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248890 | NR_002963.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248890 | AJ609460.1 | human_transcriptome |

SNORA24
Genomic Map