Assay Details
| Gene Symbol: ST7-AS2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109980.1 | 1 - 2 | 57 | 90 | |||
| GenBank mRNA | AF400042.1 | - | LIT2001 ST7OT2 |
1 - 2 | 78 | 90 | |
| CB054673.1 | - | 1 - 2 | 66 | 90 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6132 | MIR6132 | hsa-mir-6132 | -60.5kb | Anti-sense | Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38 |
| Downstream | ST7 overlapping transcript 3 | ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
91.9kb | Anti-sense | Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ST7-AS2
Genomic Map