Assay Details
| Gene Symbol: PFKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002021.2 | NP_001002021.2 | 1 - 2 | 155 | 59 | |
| XM_006724011.1 | XP_006724074.1 | 1 - 2 | 101 | 59 | ||
| XM_017028369.1 | XP_016883858.1 | 1 - 2 | 119 | 59 | ||
| XM_017028370.1 | XP_016883859.1 | 1 - 2 | 100 | 59 | ||
| GenBank mRNA | BC006422.1 | - | 1 - 2 | 101 | 4040291 | 59 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | autoimmune regulator | AIRE |
AIRE1, APECED, APS1, APSI, PGA1 |
-1.8kb | Sense | Chr. 21 - 44285838 - 44298219 on NCBI Build GRCh38 |
| Downstream | chromosome 21 open reading frame 2 | C21orf2 |
LRRC76, YF5/A2 |
24.5kb | Anti-sense | Chr. 21 - 44328944 - 44339417 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC006422.1, NM_001002021.2, XM_006724011.1, XM_017028369.1, XM_017028370.1 transcripts.
Set Membership: |
|

PFKL
Genomic Map