Assay Details
| Gene Symbol: PP7080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024158.1 | 1 - 2 | 293 | 102 | ||
| GenBank mRNA | AF370427.1 | - | 1 - 2 | 391 | 102 | |
| BC045808.1 | - | 1 - 2 | 132 | 102 | ||
| BC086861.1 | - | 1 - 2 | 274 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exocyst complex component 3 | EXOC3 |
SEC6, SEC6L1, Sec6p |
-3.2kb | Anti-sense | Chr. 5 - 443184 - 471938 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100288152 | LOC100288152 | 271.0 | Anti-sense | Chr. 5 - 473236 - 480892 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001095.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 262889 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 98469 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 149603 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 307667 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 264119 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 380362 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 297843 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 408660 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P419300 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 43804 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7909 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 44283 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P419300 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P419300 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00319011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15652764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15652764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873900 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P316046 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P419300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652771 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709889 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652769 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709882 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709897 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709899 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709885 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8110686 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110687 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8104201 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8110685 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 225458_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 225457_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015794.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015794.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015794.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015797.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015797.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015797.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015797.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015804.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015795.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015795.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015795.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015796.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015796.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015796.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015798.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015798.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015798.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015806.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015806.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015806.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015806.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015805.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015805.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015805.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015805.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015805.hg.1 | TC05001095.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169057.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169059.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169058.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000502511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | ENST00000510714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009804.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | ENST00000510604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169052.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169051.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169051.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169051.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169051.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169051.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169053.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169053.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169053.hg.1 | ENST00000342584.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169053.hg.1 | NR_024158.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169053.hg.1 | TC0500009804.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 225458_at | - | - |
| Human Genome U133 Plus 2.0 Array | 225458_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 225457_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 225457_PM_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225458_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225457_s_at10 | - | - |
| Human Genome U95 Set | 48818_s_at | - | - |
| Human Genome U95 Set | 49146_at | - | - |
| Human Genome U95 Set | 49146_PM_at | - | - |
| Human Genome U95 Set | 48818_PM_s_at | - | - |
| Human Genome U133 Set | 225457_s_at | - | - |
| Human Genome U133 Set | 225458_at | - | - |
| Human Genome U133 Set | 225458_PM_at | - | - |
| Human Genome U133 Set | 225457_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026431 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 205361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3239925 | AF370427.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3239925 | BC086861.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3239925 | NR_024158.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3239925 | AF370427.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3239925 | BC086861.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3239925 | NR_024158.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3239925 | AF370427.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3239925 | BC086861.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3239925 | NR_024158.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3239925 | AF370427.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3239925 | BC086861.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3239925 | NR_024158.1 | human_transcriptome |

PP7080
Genomic Map