| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019673.hg.1 |
ENST00000368439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019673.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019673.hg.1 |
TC01001280.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100173405.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173405.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173405.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173405.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173405.hg.1 |
TC0100010100.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000308987.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000368439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000471245.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000473344.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000474215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
ENST00000477676.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NM_001826.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010100.hg.1 |
NR_024163.1 |
Refseq |