Assay Details
| Gene Symbol: LOC105379704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036479.1 | - | 1 | 2147 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interleukin 1 receptor associated kinase 1 binding protein 1 | IRAK1BP1 |
AIP70, SIMPL |
-141.7kb | Sense | Chr. 6 - 78867486 - 78979411 on NCBI Build GRCh38 |
| Downstream | high mobility group nucleosomal binding domain 3 | HMGN3 |
PNAS-24, PNAS-25, TRIP7 |
120.8kb | Anti-sense | Chr. 6 - 79201245 - 79234738 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105379704
Genomic Map