Assay Details
| Gene Symbol: EDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036851.1 | - | 1 - 2 | 351 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | uncharacterized LOC101928309 | LOC101928309 | 268.4kb | Sense | Chr. 6 - 12582768 - 12585591 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

EDN1
Genomic Map