Assay Details
| Gene Symbol: LINC00701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038884.1 | 2 - 3 | 85 | 67 | ||
| GenBank mRNA | BC055423.1 | - | 2 - 3 | 85 | 67 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BI462068.1 | - | 90 | Chr. - - [-] on Build | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6072 | MIR6072 | hsa-mir-6072 | -224.2kb | Sense | Chr. 10 - 2076019 - 2076089 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105376351 | LOC105376351 | 115.8kb | Sense | Chr. 10 - 2446253 - 2501462 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000973.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027383 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381885 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381857 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027426 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381936 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381920 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381925 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285149 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381922 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027391 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381888 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027415 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027434 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027492 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16285134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381929 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027503 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027480 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027448 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27381877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07027395 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 174718 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 741441 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 435962 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 234872 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 939164 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 82412 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 4252 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 165753 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 407466 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 419169 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 332752 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 337210 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 116619 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 60101 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 79881 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 213743 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 243510 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 17882 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 59473 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 37535 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 306787 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 136476 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 220918 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 45349 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 202074 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3690239 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7118 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3690239 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3690239 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18810931 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18810915 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 14793 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18810931 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18810930 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18810915 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18810922 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18810915 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18810931 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10012197.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012197.hg.1 | TC10000973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012198.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012198.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012198.hg.1 | TC10000973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012199.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012199.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012199.hg.1 | TC10000973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012200.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012200.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012200.hg.1 | TC10000973.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009569.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165316.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165316.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165316.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165317.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165317.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165317.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165321.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165321.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165321.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165321.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165321.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165323.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165323.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165323.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165323.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165323.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165318.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165318.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165318.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165320.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165320.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165320.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165320.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165320.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165322.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165322.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165322.hg.1 | ENST00000418295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165322.hg.1 | NR_038884.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165322.hg.1 | TC1000009569.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042356 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042356 | - | - |

LINC00701
Genomic Map