Assay Details
| Gene Symbol: ASTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648673.1 | - | 1 | 6448 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif containing 32 | TRIM32 |
BBS11, HT2A, LGMD2H, TATIP |
-350.0kb | Anti-sense | Chr. 9 - 116687302 - 116701300 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, H/ACA box 70C |
SNORA70C | U70C | 123.1kb | Sense | Chr. 9 - 117181066 - 117181201 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ASTN2
Genomic Map