Assay Details
Gene Symbol: LOC100131174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK130404.1 | - | 1 | 1149 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MIR1-1 host gene | MIR1-1HG |
C20orf166, MIR133A2HG, dJ353C17.1 |
-4.8kb | Sense | Chr. 20 - 62550453 - 62570764 on NCBI Build GRCh38 |
Downstream | solute carrier organic anion transporter family member 4A1 | SLCO4A1 |
OATP-E, OATP1, OATP4A1, OATPE, OATPRP1, POAT, SLC21A12 |
65.1kb | Sense | Chr. 20 - 62642445 - 62690700 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
