Assay Details
| Gene Symbol: LOC646034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125175.1 | - | 1 | 2645 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger E-box binding homeobox 1 | ZEB1 |
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A |
-73.3kb | Sense | Chr. 10 - 31318495 - 31529814 on NCBI Build GRCh38 |
| Downstream | Rho GTPase activating protein 12 | ARHGAP12 | 199.5kb | Anti-sense | Chr. 10 - 31805398 - 31928876 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521417 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16308159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16308160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161323 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161311 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161391 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161302 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521418 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161314 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16308150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521403 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161307 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161348 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161347 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161294 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27521388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07161351 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 38087 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 23642 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | AF105365.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | AK024493.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | AK024497.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | AK026625.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | AL117433.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | BC007760.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | BC018982.2 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | BC098390.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 187225 | NM_006598.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 164603 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3732688 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11056 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3732688 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P18884298 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18884298 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 131203 | AL050256.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 131203 | NM_015372.1 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 4624 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18884298 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18884298 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007202.hg.1 | ENST00000420945.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3201614 | - | - |
| HumanHT-12 v4 Expression BeadChip | ILMN_3290120 | AK125175.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3201614 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_3290120 | AK125175.1 | human_transcriptome |

LOC646034
Genomic Map