Assay Details
Gene Symbol: FAM162A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK125835.1 | - | 1 | 4148 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 58 | CCDC58 | -22.3kb | Anti-sense | Chr. 3 - 122359587 - 122383281 on NCBI Build GRCh38 | |
Downstream | WD repeat domain 5B | WDR5B | 1.9kb | Anti-sense | Chr. 3 - 122411853 - 122416035 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
