Assay Details
| Gene Symbol: LOC100128517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126772.1 | - | 1 | 3646 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD83 molecule | CD83 |
BL11, HB15 |
-77.3kb | Sense | Chr. 6 - 14117256 - 14136918 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1108 | LINC01108 | LncRNA-ES1 | 62.1kb | Anti-sense | Chr. 6 - 14280127 - 14285454 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756676 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756674 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568314 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568427 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756673 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568401 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568426 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787111 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787105 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568347 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15787119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24756786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04568410 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 119419 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3239527 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23268 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3239527 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01427801 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01427801 | - | - |

LOC100128517
Genomic Map