Assay Details
Gene Symbol: LINC00322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131425.1 | - | 1 | 2446 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068763 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407332 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407327 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407248 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068634 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407219 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407305 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407255 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407342 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407228 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407231 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407256 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407241 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407275 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240032 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407221 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407242 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407257 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407322 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17239999 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407314 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407317 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407328 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407320 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407289 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407346 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407222 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407340 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407237 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407338 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407348 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407319 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240002 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407230 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407283 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407324 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407224 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407233 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407252 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407339 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407236 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407315 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068685 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17240030 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068709 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407250 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32068653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11407325 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 847366 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 417776 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22280 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416622 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 36624 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P34806979 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03593769 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03593769 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1855566 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6760717 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_95286 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1855566 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1855566 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1855566 | - | - |
