| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016706.hg.1 |
ENST00000428391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016706.hg.1 |
ENST00000428391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007054.hg.1 |
ENST00000620858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010134.hg.1 |
ENST00000502209.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007054.hg.1 |
ENST00000620858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000440461.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000629474.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000631443.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011698.hg.1 |
ENST00000632794.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016706.hg.1 |
ENST00000428391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016706.hg.1 |
ENST00000428391.1 |
Ensembl |