Assay Details
| Gene Symbol: ARPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074205.1 | - | 1 | 1151 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ARPC4-TTLL3 readthrough | ARPC4-TTLL3 | Sense | Chr. 3 - 9792548 - 9836356 on NCBI Build GRCh38 | ||
| Downstream | tubulin tyrosine ligase like 3 | TTLL3 | HOTTL | 2.9kb | Sense | Chr. 3 - 9809960 - 9836356 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ARPC4
Genomic Map