| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1558234_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011600.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011600.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011600.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011600.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17024691.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17024691.hg.1 |
TC17001833.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700194045.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700194045.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700194045.hg.1 |
TC1700011600.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700194045.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700194045.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011600.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011600.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011600.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011600.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700194044.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700194044.hg.1 |
NR_036534.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700194044.hg.1 |
TC1700011600.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700194044.hg.1 |
ENST00000590966.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700194044.hg.1 |
NR_036534.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1558234_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1558234_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1558234_at5 |
- |
- |
| Human Genome U95 Set |
46824_PM_r_at |
- |
- |
| Human Genome U95 Set |
46824_r_at |
- |
- |
| Human Genome U95 Set |
72460_at |
- |
- |
| Human Genome U95 Set |
72460_PM_at |
- |
- |