| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8167164 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8167163 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
239034_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007144.hg.1 |
ENST00000357412.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007144.hg.1 |
ENST00000624822.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007144.hg.1 |
ENST00000357412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007144.hg.1 |
ENST00000624822.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007144.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
ENST00000357412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
ENST00000624822.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
ENST00000357412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
ENST00000624822.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148697.hg.1 |
TC0X00007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00148699.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148699.hg.1 |
NR_126059.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00148699.hg.1 |
TC0X00007144.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
239034_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
239034_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
239034_at11 |
- |
- |
| Human Genome U95 Set |
64969_r_at |
- |
- |
| Human Genome U95 Set |
64969_PM_r_at |
- |
- |
| Human Genome U133 Set |
239034_at |
- |
- |
| Human Genome U133 Set |
239034_PM_at |
- |
- |