Assay Details
| Gene Symbol: LOC100131492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF370412.1 | - | 1 | 448 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein C, inactivator of coagulation factors Va and VIIIa |
PROC |
APC, PC, PROC1, THPH3, THPH4 |
-22.5kb | Anti-sense | Chr. 2 - 127418221 - 127429246 on NCBI Build GRCh38 |
| Downstream |
IWS1, SUPT6H interacting protein |
IWS1 | 26.6kb | Sense | Chr. 2 - 127480807 - 127526886 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469027 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469007 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584133 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469039 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15320096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469012 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01468850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01469028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21584143 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 161439 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 33797 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 114381 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 163434 | AB075499.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 447260 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 23329 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 241018 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 821505 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 103222 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 631112 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242238 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 701877 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 586219 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 819556 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 829229 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 539729 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 149593 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 69314 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 921796 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 908720 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 273021 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 350746 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 51177 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3420382 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22551 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00465262 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00465262 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00465262 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00465219 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00465318 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P15819852 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00465281 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35849052 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00465253 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00465281 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35849132 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00465219 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00465262 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15819852 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15819792 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00465318 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 41815 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 74012 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 116750 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 50309 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 9149 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 101618 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 33960 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 38712 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 4233 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35849194 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15819857 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465318 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15819852 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35849191 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465270 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465311 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30707773 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15819861 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465248 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465257 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465312 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465262 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35849132 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465315 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465281 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00465253 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P21854016 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35849132 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15819861 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35849208 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00465281 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15819852 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00465253 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35849191 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00465262 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00465315 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00465318 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007577.hg.1 | ENST00000619772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007577.hg.1 | ENST00000619772.1 | Ensembl |

LOC100131492
Genomic Map