Assay Details
| Gene Symbol: ST7-AS2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF400043.1 | - | LIT1938 ST7OT2 |
1 | 249 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppression of tumorigenicity 7 | ST7 |
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7 |
Anti-sense | Chr. 7 - 116953327 - 117230023 on NCBI Build GRCh38 | |
| Downstream | ST7 overlapping transcript 3 | ST7-OT3 |
NCRNA00026, ST7, ST7OT3 |
36.2kb | Anti-sense | Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |

ST7-AS2
Genomic Map