Assay Details
| Gene Symbol: LINC00654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015406.2 | 2 | 5604 | 87 | ||
| GenBank mRNA | AL137476.1 | - | 1 | 4744 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

LINC00654
Genomic Map