Assay Details
| Gene Symbol: LOC728073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL833395.1 | - | 1 | 545 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928251 | LOC101928251 | -1.1kb | Sense | Chr. 17 - 73513683 - 73518770 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100134391 | LOC100134391 | 216.4kb | Sense | Chr. 17 - 73737854 - 73757723 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457510 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457536 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457502 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041020 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040995 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041027 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17014578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457504 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31040989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31041024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10457542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17014581 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3302055 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24699 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3302055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564559_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008830.hg.1 | ENST00000583712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008830.hg.1 | ENST00000583712.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1564559_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564559_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564559_at10 | - | - |
| Human Genome U95 Set | 79357_PM_at | - | - |
| Human Genome U95 Set | 79357_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3219008 | AL833395.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3219008 | AL833395.1 | human_transcriptome |

LOC728073
Genomic Map