| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007361.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007361.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007361.hg.1 |
TC12000547.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007362.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007362.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12007362.hg.1 |
TC12000547.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158013.hg.1 |
TC1200007906.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158011.hg.1 |
TC1200007906.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
ENST00000300145.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
ENST00000551997.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
NM_033276.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200158012.hg.1 |
TC1200007906.hg.1 |
Clariom D TC |