Assay Details
| Gene Symbol: BTF3P11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026983.1 | 1 | 251 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerization domain containing 12 | KCTD12 |
C13orf2, PFET1, PFETIN |
-42.0kb | Anti-sense | Chr. 13 - 76880169 - 76886405 on NCBI Build GRCh38 |
| Downstream | aconitate decarboxylase 1 | ACOD1 |
CAD, IRG1 |
19.5kb | Sense | Chr. 13 - 76948559 - 76958642 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000290.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418703 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969935 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969986 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16650544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969983 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969933 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418689 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16650547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418726 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29418724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08969944 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 6744 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P105900 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4903 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4902 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 27907 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P107035 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P107035 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62921 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 37427 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51657 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43120 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42165 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59719 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8020 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 21756 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43040 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61824 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P105900 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3412556 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P105900 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969938 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418703 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969938 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969945 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969939 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969924 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418704 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418703 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16650547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418692 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969943 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418700 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418701 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969931 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08969932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29418693 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P107035 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P107035 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P105900 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 13988 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 4118 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P107035 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969982 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969926 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969977 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969994 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969959 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969924 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969989 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969992 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969963 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969937 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969935 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969954 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969953 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969951 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969969 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969949 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969932 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969983 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969942 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969945 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969925 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969981 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969972 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969961 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969938 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969985 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969956 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969974 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969979 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08969965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7969480 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7969481 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7969479 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 208529_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003117.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003117.hg.1 | TC13000290.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300150976.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150976.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150976.hg.1 | TC1300007545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300150973.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300150973.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150973.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300150973.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150973.hg.1 | TC1300007545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300150975.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150975.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150975.hg.1 | TC1300007545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | ENST00000426582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007545.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150974.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150974.hg.1 | NR_026983.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300150974.hg.1 | TC1300007545.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 208529_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208529_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208529_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208529_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw529172 | - | - |
| Human Genome U95 Set | 31516_f_at | - | - |
| Human Genome U95 Set | 31516_PM_f_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208529_at11 | - | - |
| Human Genome U133 Set | 208529_at | - | - |
| Human Genome U133 Set | 208529_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050313 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000046 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A000046 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 133238 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309354 | NR_026983.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309354 | NR_026983.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309354 | NR_026983.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309354 | NR_026983.1 | human_transcriptome |

BTF3P11
Genomic Map