| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000421538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000444890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000444890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000444890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000444890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000444890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
ENST00000609737.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008999.hg.1 |
NR_027036.1 |
Refseq |