Assay Details
| Gene Symbol: DLG5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024585.1 | 1 | 1342 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | discs large MAGUK scaffold protein 5 | DLG5 |
LP-DLG, P-DLG5, PDLG |
-222.0 | Anti-sense | Chr. 10 - 77790787 - 77926590 on NCBI Build GRCh38 |
| Downstream | RNA polymerase III subunit A | POLR3A |
ADDH, HLD7, RPC1, RPC155, hRPC155 |
45.3kb | Anti-sense | Chr. 10 - 77975149 - 78029540 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000557.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 155009 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 277740 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3287710 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4645 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4646 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62113 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18987555 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P18987555 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3287710 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3287710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16358439 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336560 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16358439 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336560 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07336576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27720202 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P918786 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16358439 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336598 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336571 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336592 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336595 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336601 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336566 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336603 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336578 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336590 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16358437 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336587 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16358440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16358442 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07336559 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18987555 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7928546 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7928545 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1562598_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 239565_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11754480_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11754480_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005961.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005961.hg.1 | TC10000557.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005959.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005959.hg.1 | TC10000557.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005960.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005960.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005960.hg.1 | TC10000557.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005962.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005962.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005962.hg.1 | TC10000557.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000155233.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155233.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155233.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155233.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155233.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155236.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155236.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155236.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155236.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155236.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008152.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155235.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155235.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155235.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155235.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155235.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155231.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155231.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155231.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155237.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155232.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155232.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155232.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | ENST00000449852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | ENST00000632919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | NR_024585.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155238.hg.1 | TC1000008152.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1562598_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562598_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562598_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044156 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3249338 | NR_024585.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3249338 | AK094329.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3249338 | BM981914.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249338 | NR_024585.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249338 | AK094329.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3249338 | BM981914.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249338 | NR_024585.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249338 | AK094329.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3249338 | BM981914.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249338 | NR_024585.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249338 | AK094329.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3249338 | BM981914.1 | human_transcriptome |

DLG5-AS1
Genomic Map