Assay Details
| Gene Symbol: HIPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF111850.2 | - | 1 | 503 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | C-type lectin domain family 2 member L | CLEC2L | -67.0kb | Anti-sense | Chr. 7 - 139523928 - 139544985 on NCBI Build GRCh38 | |
| Downstream | thromboxane A synthase 1 | TBXAS1 |
BDPLT14, CYP5, CYP5A1, GHOSAL, THAS, TS, TXAS, TXS |
163.6kb | Anti-sense | Chr. 7 - 139778248 - 140020325 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

HIPK2
Genomic Map