Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF086356.1 | - | 1 | 252 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | low density lipoprotein receptor class A domain containing 1 | LDLRAD1 | -21.6kb | Sense | Chr. 1 - 54007298 - 54018186 on NCBI Build GRCh38 | |
| Downstream | transcription elongation factor A N-terminal and central domain containing 2 | TCEANC2 | C1orf83 | 13.4kb | Anti-sense | Chr. 1 - 54053572 - 54112519 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMEM59
Genomic Map