Assay Details
| Gene Symbol: MYL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006471.3 | NP_006462.1 | 1 - 2 | 383 | 70 | |
| GenBank mRNA | AK222569.1 | - | 1 - 2 | 59 | 70 | |
| AK291145.1 | - | 1 - 2 | 79 | 70 | ||
| BC031972.1 | - | 1 - 2 | 383 | 4836883 | 70 | |
| BC032748.1 | - | 1 - 2 | 54 | 5563221 | 70 | |
| D82059.1 | - | 1 - 2 | 49 | 70 | ||
| EU794621.1 | - | 1 - 2 | 383 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000016.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291145.1,BC031972.1,BC032748.1,D82059.1,EU794621.1,NM_006471.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000172.hg.1 | ENST00000217652.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000172.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000172.hg.1 | TC18000016.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000173.hg.1 | ENST00000217652.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000173.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000173.hg.1 | TC18000016.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144570.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144566.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000577510.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000580353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000577510.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000580353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144563.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000577510.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000580353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000583449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000577510.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000578562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000580353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | ENST00000583449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144562.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000578038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000578038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144569.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000578038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_006471.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | TC1800006506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000217652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000536605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000578038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000578611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000579226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | ENST00000580887.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303047.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_001303049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144568.hg.1 | NM_006471.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 184808 | - | - |

MYL12A
Genomic Map