Assay Details
| Gene Symbol: SYDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006710946.3 | XP_006711009.1 | 5 | 5055 | 91 | |
| GenBank mRNA | AK075186.1 | - | 1 | 2242 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4423 | MIR4423 | mir-4423 | -40.4kb | Anti-sense | Chr. 1 - 85133794 - 85133873 on NCBI Build GRCh38 |
| Downstream | chromosome 1 open reading frame 52 | C1orf52 | gm117 | 73.1kb | Sense | Chr. 1 - 85249953 - 85259672 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SYDE2
Genomic Map