Assay Details
| Gene Symbol: LOC105374895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK092525.1 | - | 1 - 2 | 383 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724096 | LOC102724096 | -301.8kb | Anti-sense | Chr. 6 - 4186320 - 4190029 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105374896 | LOC105374896 | 104.9kb | Anti-sense | Chr. 6 - 4599288 - 4602420 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374895
Genomic Map