Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
238456_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006839.hg.1 |
TC05000480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006839.hg.1 |
ENST00000513175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006839.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006838.hg.1 |
TC05000480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006838.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006840.hg.1 |
TC05000480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05006840.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193696.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193696.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193696.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193698.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193698.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193698.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193695.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193695.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193695.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193689.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193689.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193689.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193692.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193692.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193692.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193687.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193687.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193687.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193691.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193691.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193691.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193693.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193693.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193693.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193697.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193697.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193697.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193690.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193690.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193690.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500193701.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193701.hg.1 |
NR_036530.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193701.hg.1 |
TC0500013209.hg.1 |
Clariom D TC |
Human Genome U95 Set |
68718_at |
- |
- |
Human Genome U95 Set |
68718_PM_at |
- |
- |
Human Genome U95 Set |
84118_at |
- |
- |
Human Genome U95 Set |
84118_PM_at |
- |
- |
Human Genome U133 Set |
238456_at |
- |
- |
Human Genome U133 Set |
238456_PM_at |
- |
- |