Assay Details
| Gene Symbol: EBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022659.3 | NP_073150.2 | 16 | 4224 | 71 | |
| GenBank mRNA | AK093878.1 | - | 1 | 1549 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle associated 2 | CDCA2 |
PPP1R81, Repo-Man |
-330.4kb | Anti-sense | Chr. 8 - 25458569 - 25511308 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100507236 | LOC100507236 | 97.3kb | Sense | Chr. 8 - 25941229 - 25943863 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EBF2
Genomic Map