Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurexophilin and PC-esterase domain family member 2 | NXPE2 | FAM55B | -499.7kb | Anti-sense | Chr. 11 - 114464276 - 114813702 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105369509 | LOC105369509 | 122.7kb | Anti-sense | Chr. 11 - 115333577 - 115340439 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map