Assay Details
| Gene Symbol: LOC100506444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125918.1 | 2 - 3 | 443 | 100 | ||
| GenBank mRNA | AK129929.1 | - | 2 - 3 | 443 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ligand of numb-protein X 1 | LNX1 |
LNX, MPDZ, PDZRN2 |
-64.3kb | Anti-sense | Chr. 4 - 53458264 - 53652524 on NCBI Build GRCh38 |
| Downstream | ribosomal protein L21 pseudogene 44 | RPL21P44 | RPL21_17_477 | 251.1kb | Anti-sense | Chr. 4 - 53985486 - 53987282 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102459 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102409 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102460 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256605 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102448 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102449 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102451 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256614 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256682 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102417 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256588 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256700 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256586 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102416 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102465 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102431 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256574 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256650 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256643 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102508 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102424 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256685 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102463 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256739 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102425 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102410 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256661 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256604 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102427 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256660 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15567473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102476 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23256630 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15567472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03102354 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | ENST00000508112.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007528.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151518.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400151518.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151518.hg.1 | ENST00000506950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400151518.hg.1 | NR_125918.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400151518.hg.1 | TC0400007528.hg.1 | Clariom D TC |

LOC100506444
Genomic Map