Assay Details
| Gene Symbol: PLA2G4E-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120334.1 | 3 - 4 | 546 | 74 | ||
| GenBank mRNA | BC043275.2 | - | 3 - 4 | 545 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phospholipase A2 group IVE | PLA2G4E | Anti-sense | Chr. 15 - 41981582 - 42050703 on NCBI Build GRCh38 | ||
| Downstream | phospholipase A2 group IVD | PLA2G4D | cPLA2delta | 68.5kb | Anti-sense | Chr. 15 - 42067562 - 42094559 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007019.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149374.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149374.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149374.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149374.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149374.hg.1 | TC1500007019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500149376.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149376.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149376.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149376.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149376.hg.1 | TC1500007019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500149372.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149372.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149372.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149372.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149372.hg.1 | TC1500007019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500149369.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149369.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149369.hg.1 | ENST00000499478.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500149369.hg.1 | NR_120334.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500149369.hg.1 | TC1500007019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |

PLA2G4E-AS1
Genomic Map